Learn More
Medchemexpress LLC Cholesterol 24-hydroxylase-IN-1 | 1613480-70-8 | MFCD29913759 | >99.55% | C17H23N5O | 50 MG
SDP
Catalog No. 5000354274
Encompass_Preferred
Cholesterol 24-hydroxylase-IN-1 (compound 17) is a potent, orally active, and highly selective cholesterol 24-hydroxylase (CH24H or CYP46A1) inhibitor with an IC50 of 8.5 nM. It can cross the blood-brain barrier.
  • Potent, orally active, and highly selective inhibitor
  • Inhibits cholesterol 24-hydroxylase (CH24H or CYP46A1) with an IC50 of 8.5 nM
  • Can cross the blood-brain barrier
Catalog No. 50-003-54274 Supplier Medchemexpress LLC Supplier No. HY14430950MG
Cholesterol 24-hydroxylase-IN-1 (compound 17) is a potent, orally active, and highly selective cholesterol 24-hydroxylase (CH24H or CYP46A1) inhibitor with an IC50 of 8.5 nM. It can cross the blood-brain barrier.
  • Potent, orally active, and highly selective inhibitor
  • Inhibits cholesterol 24-hydroxylase (CH24H or CYP46A1) with an IC50 of 8.5 nM
  • Can cross the blood-brain barrier