Learn More
Enzo Life Sciences Cholesterol 3-sulfate. sodium salt (5mg). CAS: 2864-50-8
SDP
Supplier:  Enzo Life Sciences ALX370009M005
Encompass_Preferred
Activates multiple protein kinase C (PKC) iso-enzymes, especially the ε, η and ζ isoforms. Purity: One spot by TLC. Appearance: White to off-white powder. Solubility: Soluble in methanol. Long Term Storage: +4°C.
Catalog No. 50-200-8889