Learn More
American Radiolabeled Chemicals Inc CHOLESTEROL-4-14C 250UC
SDP
Catalog No. 5090106230
Encompass_Preferred
Cholesterol-4-14C 250uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 386.6
Catalog No. 50-901-06230 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 0857250UC
Add to Cart
Add to Cart
Request a Quote
Cholesterol-4-14C 250uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 386.6