Learn More
Avanti Polar Lipids CHOLESTEROL 500MG
Supplier:  Avanti Polar Lipids 700000P500MG
Encompass
Cholesterol, Purity: >98%, HPLC, White Powder, Molecular Weight: 386.66, Molecular Formula: C27H46O, Percent Composition: C 83.87% H 11.99% O 4.14%
Catalog No. NC9185980