Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol, 95%, Thermo Scientific Chemicals

Catalog No. AAA1147030
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1147030 250 g
AAA1147018 50 g
AAA114700B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1147030 Supplier Thermo Scientific Chemicals Supplier No. A1147030
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

A laboratory purified preparation of the animal sterol, Cholesterol, CAS # 57-88-5, is available in powder form for experimental use.

Cholesterol is used in the preparation of liposome. It is used to control the functions of transporters and coordinating cell proteins present in plasma membrane. It acts as a part of cell membranes. It benefits in rapid diagnostics. It is a component of all biological membranes and inhibits basal channel activity in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
  • Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
  • A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
  • It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems

Applications

  • Suitable for in vitro and in vivo applications
  • Stabilized in powder form for laboratory use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00003646
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
Synonym cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
PubChem CID 5997
ChEBI CHEBI:16113
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 147°C to 150°C
Density 1.067
Boiling Point 360°C
Assay Percent Range 95%
Quantity 250 g
Beilstein 1915888
Merck Index 14,2201
Solubility Information Soluble in alcohols,ether,chloroform,petroleum ether,pyridine,isopropyl myristate,acetone,ethylacetate,benzene,fats,oils and dioxane.
Optical Rotation −36° (c=2 in dioxane)
Formula Weight 386.66
Percent Purity 95%
Chemical Name or Material Cholesterol
Show More Show Less
EINECSNumber 200-353-2
RTECSNumber FZ8400000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.