Learn More
Medchemexpress LLC Cholesterol β-D-glucoside | 7073-61-2 | 99.4% | 548.79 g/mol | C33H56O6 | 5 MG
SDP
Catalog No. 5000263064
Encompass_Preferred
Cholesterol β-D-glucoside is a small-molecule activator of heat shock transcription factor 1 (HSF1) used in research applications; it has been reported to induce HSP70 expression in fibroblast cells and is supplied as a high-purity powder.
  • Activates HSF1 and induces HSP70 in fibroblast cells.
  • High purity: 99.4% as stated on the certificate of analysis.
  • Molecular formula C33H56O6 and molecular weight 548.79 g/mol.
  • Supplied as a powder in small research quantities, including 1 mg, 5 mg, and 10 mg packages.
  • Powder storage: -20 °C, stable for up to 3 years; in solvent: -80 °C for 6 months or -20 °C for 1 month.
  • Intended for research use only.
Catalog No. 50-002-63064 Supplier Medchemexpress LLC Supplier No. HY1339615MG
Cholesterol β-D-glucoside is a small-molecule activator of heat shock transcription factor 1 (HSF1) used in research applications; it has been reported to induce HSP70 expression in fibroblast cells and is supplied as a high-purity powder.
  • Activates HSF1 and induces HSP70 in fibroblast cells.
  • High purity: 99.4% as stated on the certificate of analysis.
  • Molecular formula C33H56O6 and molecular weight 548.79 g/mol.
  • Supplied as a powder in small research quantities, including 1 mg, 5 mg, and 10 mg packages.
  • Powder storage: -20 °C, stable for up to 3 years; in solvent: -80 °C for 6 months or -20 °C for 1 month.
  • Intended for research use only.