Learn More
Medchemexpress LLC Cholesterol β-D-glucoside | 7073-61-2 | MFCD00056433 | 99.4% | C33H56O6 | 10MG
SDP
Catalog No. 5000259201
Encompass_Preferred
Cholesterol β-D-glucoside is a steroid glycoside used as a research reagent to activate heat shock transcription factor 1 (HSF1) and induce HSP70 expression in fibroblast cells. It is provided as a high-purity solid for cell biology and biochemical studies of heat-shock response pathways.
  • Activates heat shock transcription factor 1 (HSF1) and induces HSP70.
  • High purity (~99.4%) for reliable experimental results.
  • Molecular formula C33H56O6; molecular weight 548.79.
  • Solid, white to off-white appearance suitable for powder storage.
  • Stable when stored at -20°C as powder (3 years) and in solvent at -80°C (6 months).
  • Certificate of analysis and safety data sheet are available.
Catalog No. 50-002-59201 Supplier Medchemexpress LLC Supplier No. HY13396110MG
Cholesterol β-D-glucoside is a steroid glycoside used as a research reagent to activate heat shock transcription factor 1 (HSF1) and induce HSP70 expression in fibroblast cells. It is provided as a high-purity solid for cell biology and biochemical studies of heat-shock response pathways.
  • Activates heat shock transcription factor 1 (HSF1) and induces HSP70.
  • High purity (~99.4%) for reliable experimental results.
  • Molecular formula C33H56O6; molecular weight 548.79.
  • Solid, white to off-white appearance suitable for powder storage.
  • Stable when stored at -20°C as powder (3 years) and in solvent at -80°C (6 months).
  • Certificate of analysis and safety data sheet are available.