Learn More
Medchemexpress LLC Cholesterol-d6-1 | 92543-08-3 | 99.82% | 392.69 | 1 MG
SDP
Catalog No. 5000427414
Encompass_Preferred
Cholesterol-d6-1 is deuterium labeled Cholesterol, a primary sterol in mammals that makes up 20-25% of the plasma membrane's structural component. It plays a significant role in determining the fluidity and permeability characteristics of the membrane, as well as the function of transporters and signaling proteins. It also acts as an endogenous estrogen-related receptor α (ERRα) agonist.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-27414 Supplier Medchemexpress LLC Supplier No. HYN0322S21MG
Add to Cart
Add to Cart
Cholesterol-d6-1 is deuterium labeled Cholesterol, a primary sterol in mammals that makes up 20-25% of the plasma membrane's structural component. It plays a significant role in determining the fluidity and permeability characteristics of the membrane, as well as the function of transporters and signaling proteins. It also acts as an endogenous estrogen-related receptor α (ERRα) agonist.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS