Learn More
Cayman Chemical Cholesterol-d6 10mg
SDP
Catalog No. 503015740
Encompass_Preferred
An internal standard for the quantification of cholesterol by GC- or LC-MS
Catalog No. 50-301-5740 Supplier Cayman Chemical Supplier No. 2554610 MG
An internal standard for the quantification of cholesterol by GC- or LC-MS