Learn More
Cayman Chemical Cholesterol-DoxorubIcIn 10mg
SDP
Catalog No. 502894559
Encompass_Preferred
A prodrug form of doxorubicin; reduces MCF-7 and MDA-MB-231 breast cancer cell viability when combined with TPGS in nanoassemblies
Catalog No. 50-289-4559 Supplier Cayman Chemical Supplier No. 900419810 MG
A prodrug form of doxorubicin; reduces MCF-7 and MDA-MB-231 breast cancer cell viability when combined with TPGS in nanoassemblies