Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol, NF, 95-102%, Spectrum™ Chemical
SDP

Catalog No. 18601652 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-652 25 g Amber Glass Bottle
18-601-653 125 g Amber Glass Bottle
18-601-654 500 g Amber Glass Bottle
18-601-655 2.5 kg Amber Glass Bottle
18-601-656 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-652 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH13525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CH135, 57-88-5, C27H46O

Spectrum™ Chemical Cholesterol, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.66
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
IUPAC Name (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Melting Range 147-150°C
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -34° to -38°
Percent Purity 95-102%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Cholesterol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.