Learn More
MOLECULAR DEPOT LLC Cholesterol Oxidase - 250 U
Catalog No. 5000013357
Encompass_Preferred
Cholesterol Oxidase (250 U) consists of a highly purified powdered Cholesterol Oxidase enzyme a flavoprotein involved in the first step of cholesterol catabolism The enzyme first oxidizes cholesterol to cholest-5-en-3-one then isomerizes it to cholest-4-en-3-one while producing hydrogen peroxide For in vitro research use only
Catalog No. 50-000-13357 Supplier MOLECULAR DEPOT LLC Supplier No. P2010002
Cholesterol Oxidase (250 U) consists of a highly purified powdered Cholesterol Oxidase enzyme a flavoprotein involved in the first step of cholesterol catabolism The enzyme first oxidizes cholesterol to cholest-5-en-3-one then isomerizes it to cholest-4-en-3-one while producing hydrogen peroxide For in vitro research use only