Learn More
Medchemexpress LLC Cholesterol-PEG 600 | 69068-97-9 | 1,055.60 g/mol | C68H110O8 | 1 G
SDP
Catalog No. 5000283102
Encompass_Preferred
Cholesterol-PEG 600 is a cholesterol-terminated polyethylene glycol reagent (average PEG MW ~600) used in life-science research to modify lipid assemblies and improve amphiphilic conjugation. It is supplied as an oil (off-white to light yellow) for formulation into liposomes, nanoparticles, and other delivery systems. For research use only.
Catalog No. 50-002-83102 Supplier Medchemexpress LLC Supplier No. HYW1275581G
Cholesterol-PEG 600 is a cholesterol-terminated polyethylene glycol reagent (average PEG MW ~600) used in life-science research to modify lipid assemblies and improve amphiphilic conjugation. It is supplied as an oil (off-white to light yellow) for formulation into liposomes, nanoparticles, and other delivery systems. For research use only.