Learn More
Kyfora Bio Cholesterol -PEG 600 25 MG
Catalog No. 502559790
Encompass_Preferred
Cholesterol-PEG 600 is a PEGylated cholesterol derivative designed for use in lipid nanoparticle (LNP) formulation and drug delivery applications. Cholesterol serves as a structural component in LNPs, enhancing membrane integrity and stability. The PEGylation of cholesterol-PEG 600 adds a protective surface layer to the LNP, reducing opsonization and improving circulation time in vivo, which is essential for effective delivery of therapeutic agents. For research use only.Key Applications: Lipid nanoparticles (LNPs), drug delivery
Catalog No. 50-255-9790 Supplier Kyfora Bio Supplier No. 1454325
Cholesterol-PEG 600 is a PEGylated cholesterol derivative designed for use in lipid nanoparticle (LNP) formulation and drug delivery applications. Cholesterol serves as a structural component in LNPs, enhancing membrane integrity and stability. The PEGylation of cholesterol-PEG 600 adds a protective surface layer to the LNP, reducing opsonization and improving circulation time in vivo, which is essential for effective delivery of therapeutic agents. For research use only.Key Applications: Lipid nanoparticles (LNPs), drug delivery