Learn More
Medchemexpress LLC Cholesterol-PEG-azide | 85.7% | 3400 (average) | 5 MG
SDP
Catalog No. 5000280985
Encompass_Preferred
Cholesterol-PEG-azide (average MW 3400) is an amphiphilic cholesterol-polyethylene glycol conjugate bearing a terminal azide functional group. It serves as a clickable lipid anchor for bioconjugation and can self-assemble into micelles for use in drug delivery research and nanoparticle surface modification. Supplied as a white to off-white solid for research use.
  • Provides an azide functional group for click chemistry.
  • Amphiphilic structure enables micelle formation and membrane anchoring.
  • Average PEG spacer (MW ~3400) offers flexible linker length.
  • Suitable for drug delivery and nanoparticle surface modification studies.
  • Stable when stored under recommended conditions (-20°C, protected from light).
Catalog No. 50-002-80985 Supplier Medchemexpress LLC Supplier No. HYW4406955MG
Cholesterol-PEG-azide (average MW 3400) is an amphiphilic cholesterol-polyethylene glycol conjugate bearing a terminal azide functional group. It serves as a clickable lipid anchor for bioconjugation and can self-assemble into micelles for use in drug delivery research and nanoparticle surface modification. Supplied as a white to off-white solid for research use.
  • Provides an azide functional group for click chemistry.
  • Amphiphilic structure enables micelle formation and membrane anchoring.
  • Average PEG spacer (MW ~3400) offers flexible linker length.
  • Suitable for drug delivery and nanoparticle surface modification studies.
  • Stable when stored under recommended conditions (-20°C, protected from light).