Learn More
Medchemexpress LLC Cholesterol-PEG-biotin | 2000 (average) | (C2H4O)nC40H66N4O4S | 50 MG
SDP
Catalog No. 5000280670
Encompass_Preferred
Cholesterol-PEG-Biotin (MW 2000) is a PEGylated lipid conjugate combining a cholesterol anchor with a biotin tag via an approximate 2,000 Da PEG spacer. It is designed to incorporate into lipid bilayers or liposomes and present biotin for high-affinity binding to avidin or streptavidin, enabling surface immobilization, targeted capture, and bioconjugation in biochemical and nanotechnology applications. Supplied as a powder with low-temperature storage recommendations.
  • Provides a biotin handle for strong avidin/streptavidin binding.
  • Anchors to lipid membranes via a cholesterol moiety.
  • Peg spacer increases aqueous solubility and reduces steric hindrance.
  • Enables liposome functionalization and surface immobilization.
  • Supplied as a powder with recommended low-temperature storage.
Catalog No. 50-002-80670 Supplier Medchemexpress LLC Supplier No. HYW44071150MG
Cholesterol-PEG-Biotin (MW 2000) is a PEGylated lipid conjugate combining a cholesterol anchor with a biotin tag via an approximate 2,000 Da PEG spacer. It is designed to incorporate into lipid bilayers or liposomes and present biotin for high-affinity binding to avidin or streptavidin, enabling surface immobilization, targeted capture, and bioconjugation in biochemical and nanotechnology applications. Supplied as a powder with low-temperature storage recommendations.
  • Provides a biotin handle for strong avidin/streptavidin binding.
  • Anchors to lipid membranes via a cholesterol moiety.
  • Peg spacer increases aqueous solubility and reduces steric hindrance.
  • Enables liposome functionalization and surface immobilization.
  • Supplied as a powder with recommended low-temperature storage.