Learn More
Medchemexpress LLC Cholesterol-PEG-biotin | 2000 (average) Da | 25 MG
SDP
Catalog No. 5000281255
Encompass_Preferred
Cholesterol-PEG-Biotin (MW 2000) is a biotin-conjugated, PEGylated cholesterol lipid designed as a membrane anchor for liposomes and other membrane-mimetic systems. The PEG(2000) spacer increases aqueous solubility and provides steric stabilization, while the terminal biotin enables strong, specific capture by avidin or streptavidin. Supplied as a powder for research use.
Catalog No. 50-002-81255 Supplier Medchemexpress LLC Supplier No. HYW44071125MG
Cholesterol-PEG-Biotin (MW 2000) is a biotin-conjugated, PEGylated cholesterol lipid designed as a membrane anchor for liposomes and other membrane-mimetic systems. The PEG(2000) spacer increases aqueous solubility and provides steric stabilization, while the terminal biotin enables strong, specific capture by avidin or streptavidin. Supplied as a powder for research use.