Learn More
Medchemexpress LLC Cholesterol-PEG-MAL | 95.1% | 2000 (average) | 100 MG
SDP
Catalog No. 5000281637
Encompass_Preferred
Cholesterol-PEG-MAL (MW 2000) is a cholesterol-terminated polyethylene glycol bearing a terminal maleimide group. It serves as a pegylated lipid for liposome and nanoparticle preparation, enabling membrane anchoring and covalent conjugation to thiol-containing molecules. Supplied as a light yellow solid, it is soluble in DMSO and intended for research use.
  • enables thiol-specific covalent conjugation via maleimide chemistry.
  • provides a hydrophobic anchor for stable membrane insertion.
  • average molecular weight 2000 for predictable spacer length.
  • high purity suitable for research applications.
  • soluble in DMSO at high concentration with sonication.
  • stable when stored frozen as a powder.
Catalog No. 50-002-81637 Supplier Medchemexpress LLC Supplier No. HYW440719100MG
Cholesterol-PEG-MAL (MW 2000) is a cholesterol-terminated polyethylene glycol bearing a terminal maleimide group. It serves as a pegylated lipid for liposome and nanoparticle preparation, enabling membrane anchoring and covalent conjugation to thiol-containing molecules. Supplied as a light yellow solid, it is soluble in DMSO and intended for research use.
  • enables thiol-specific covalent conjugation via maleimide chemistry.
  • provides a hydrophobic anchor for stable membrane insertion.
  • average molecular weight 2000 for predictable spacer length.
  • high purity suitable for research applications.
  • soluble in DMSO at high concentration with sonication.
  • stable when stored frozen as a powder.