Learn More
Medchemexpress LLC Cholesterol-PEG-MAL | 95.1% | 2000 (average) | 5 MG
SDP
Catalog No. 5000281257
Encompass_Preferred
Cholesterol-PEG-MAL (MW 2000) is a PEGylated cholesterol derivative bearing a terminal maleimide group. It self-assembles in aqueous media and is used to form liposomes, micelles, and nanoparticles, and to conjugate thiol-containing molecules via maleimide-thiol chemistry. Intended for research use only.
  • Reactive maleimide end group enables stable thiol conjugation.
  • Amphiphilic cholesterol-PEG structure promotes self-assembly into liposomes and micelles.
  • Average molecular weight ~2000 provides steric stabilization in formulations.
  • Available in small research quantities suitable for formulation development.
  • Documentation available for quality verification and handling.
Catalog No. 50-002-81257 Supplier Medchemexpress LLC Supplier No. HYW4407195MG
Cholesterol-PEG-MAL (MW 2000) is a PEGylated cholesterol derivative bearing a terminal maleimide group. It self-assembles in aqueous media and is used to form liposomes, micelles, and nanoparticles, and to conjugate thiol-containing molecules via maleimide-thiol chemistry. Intended for research use only.
  • Reactive maleimide end group enables stable thiol conjugation.
  • Amphiphilic cholesterol-PEG structure promotes self-assembly into liposomes and micelles.
  • Average molecular weight ~2000 provides steric stabilization in formulations.
  • Available in small research quantities suitable for formulation development.
  • Documentation available for quality verification and handling.