Learn More
Medchemexpress LLC Cholesterol-PEG-maleimide | 95.1% | 2000 Da (average) | 10 MG
SDP
Catalog No. 5000280496
Encompass_Preferred
Cholesterol-PEG-maleimide (average MW 2000) is a cholesterol-terminated polyethylene glycol conjugate bearing a maleimide reactive group. It self-assembles in aqueous media and is used to modify liposomes or nanoparticles and to covalently attach thiol-containing ligands via maleimide-thiol chemistry.
  • Maleimide functional group reacts selectively with thiol groups to form stable thioether bonds.
  • Cholesterol terminus anchors into lipid bilayers for stable surface modification.
  • PEG (average MW 2000) provides steric stabilization and improved circulation time.
  • Amphiphilic structure facilitates liposome and nanoparticle formulation.
  • Available in small research pack sizes suitable for formulation and conjugation studies.
Catalog No. 50-002-80496 Supplier Medchemexpress LLC Supplier No. HYW44071910MG
Cholesterol-PEG-maleimide (average MW 2000) is a cholesterol-terminated polyethylene glycol conjugate bearing a maleimide reactive group. It self-assembles in aqueous media and is used to modify liposomes or nanoparticles and to covalently attach thiol-containing ligands via maleimide-thiol chemistry.
  • Maleimide functional group reacts selectively with thiol groups to form stable thioether bonds.
  • Cholesterol terminus anchors into lipid bilayers for stable surface modification.
  • PEG (average MW 2000) provides steric stabilization and improved circulation time.
  • Amphiphilic structure facilitates liposome and nanoparticle formulation.
  • Available in small research pack sizes suitable for formulation and conjugation studies.