Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014734 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4734 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4734 Supplier MilliporeSigma Supelco Supplier No. PHR1533500MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-88-5
Density 1.067 g/mL (at 25°C)
Physical Form Powder
Linear Formula C27H46 O
MDL Number MFCD00003646
Melting Point 147°C to 149°C
Boiling Point 360°C
Synonym Cholesterol; 3 beta-Hydroxy-5-cholestene; 5-Cholesten-3 beta-ol
Solubility Soluble in water
Type Certified Reference Material
Quantity 500 mg
Formula Weight 386.65
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.