Learn More
Sigma Aldrich Fine Chemicals Biosciences Cholesterol Quantification
Catalog No. 502905159
Encompass
Cholesterol is a lipid that constitutes 20-25% of the structural components of cell membranes. Cholesterol presence assure membrane structural fluidity. Cholesterol also plays a role as an intermediate in several hormones and bile acid biosynthesis and vitamin D. Cholesterol also regulates the functions of the transporters and signaling proteins present on the plasma membrane. The major sites of cholesterol synthesis are the small intestine and liver. Cholesterol circulates in the blood as both the free acid and as cholesterol esters. Increased cholesterol levels are associated with the development of atherosclerosis and cardiovascular diseases. Thus serum cholesterol regulation has an important therapeutic role.The Cholesterol Quantification Assay Kit provides a simple and quick procedure for detecting and measuring very low concentrations of Cholesterol. This kit can measure free cholesterol cholesteryl esters or total cholesterol (which constitutes both free cholesterol and chole
Catalog No. 50-290-5159 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CS00051KT
Add to Cart
Add to Cart
Cholesterol is a lipid that constitutes 20-25% of the structural components of cell membranes. Cholesterol presence assure membrane structural fluidity. Cholesterol also plays a role as an intermediate in several hormones and bile acid biosynthesis and vitamin D. Cholesterol also regulates the functions of the transporters and signaling proteins present on the plasma membrane. The major sites of cholesterol synthesis are the small intestine and liver. Cholesterol circulates in the blood as both the free acid and as cholesterol esters. Increased cholesterol levels are associated with the development of atherosclerosis and cardiovascular diseases. Thus serum cholesterol regulation has an important therapeutic role.The Cholesterol Quantification Assay Kit provides a simple and quick procedure for detecting and measuring very low concentrations of Cholesterol. This kit can measure free cholesterol cholesteryl esters or total cholesterol (which constitutes both free cholesterol and chole