Learn More
Medchemexpress LLC Cholesterol-SA-PEG-Azide (MW 2000) | 90.0% | 2000 (Average) | 25 MG
SDP
Catalog No. 5000384409
Encompass_Preferred
Cholesterol-SA-PEG-Azide (MW 2000) is a lipid designed for drug delivery. It incorporates Cholesterol, Succinic acid, PEG units, and an azide group. The azide group allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing an alkyne group.
  • Used for drug delivery
  • Contains cholesterol, succinic acid, PEG units, and an azide group
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition reaction with alkyne-containing molecules
Catalog No. 50-003-84409 Supplier Medchemexpress LLC Supplier No. 25MGHY171562
Cholesterol-SA-PEG-Azide (MW 2000) is a lipid designed for drug delivery. It incorporates Cholesterol, Succinic acid, PEG units, and an azide group. The azide group allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing an alkyne group.
  • Used for drug delivery
  • Contains cholesterol, succinic acid, PEG units, and an azide group
  • Azide group enables copper-catalyzed azide-alkyne cycloaddition reaction with alkyne-containing molecules