Learn More
Medchemexpress LLC Cholesterol stearate-d6 | 2692624-28-3 | 99.1% | 659.15 | 1 MG
SDP
Catalog No. 5000451865
Encompass_Preferred
Cholesterol stearate-d6 is a deuterium-labeled Cholesterol stearate primarily intended for research use. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-51865 Supplier Medchemexpress LLC Supplier No. HYW010695S1MG
Cholesterol stearate-d6 is a deuterium-labeled Cholesterol stearate primarily intended for research use. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for analysis by NMR, GC-MS, or LC-MS