Learn More
Medchemexpress LLC Cholesterol stearate-d6 | 2692624-28-3 | 99.1% | 659.15 | 10 MG
SDP
Catalog No. 5000451774
Encompass_Preferred
Cholesterol stearate-d6 is the deuterium labeled Cholesterol stearate. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-51774 Supplier Medchemexpress LLC Supplier No. HYW010695S10MG
Cholesterol stearate-d6 is the deuterium labeled Cholesterol stearate. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS