Learn More
Kyfora Bio Cholesterol Sulfate, sodium salt 25 MG
Catalog No. 502559791
Encompass_Preferred
Cholesterol Sulfate (sodium salt) is a naturally occurring sterol sulfate found in cell membranes, where it plays a role in lipid metabolism, membrane stabilization, and serves as a precursor in steroid biosynthesis. This compound is utilized in the preparation of micelles and in the formulation of liposomes, offering insights into membrane properties and dynamics in cell membrane studies. For research use only.Key Applications: Micelles, liposomes
Catalog No. 50-255-9791 Supplier Kyfora Bio Supplier No. 1453525
Cholesterol Sulfate (sodium salt) is a naturally occurring sterol sulfate found in cell membranes, where it plays a role in lipid metabolism, membrane stabilization, and serves as a precursor in steroid biosynthesis. This compound is utilized in the preparation of micelles and in the formulation of liposomes, offering insights into membrane properties and dynamics in cell membrane studies. For research use only.Key Applications: Micelles, liposomes