Learn More
Cayman Chemical Cholesterol Sulfate (sodium salt), Purity: ≥95%, CAS: 2864-50-8, 250 mg
SDP
Catalog No. 502972409
Encompass_Preferred
An endogenous component of cell membranes where it serves as a substrate for steroid synthesis, influences lipid metabolism, stabilizes cell membranes, regulates the activity of functional proteins, and plays a role in keratinocyte differentiation and development of the epidermal barrier
Catalog No. 50-297-2409 Supplier Cayman Chemical Supplier No. 15106250 MG
An endogenous component of cell membranes where it serves as a substrate for steroid synthesis, influences lipid metabolism, stabilizes cell membranes, regulates the activity of functional proteins, and plays a role in keratinocyte differentiation and development of the epidermal barrier