Learn More
Medchemexpress LLC Cholesterol (Water Soluble) | 98.0% | 5 MG
SDP
Catalog No. 5000427309
Encompass_Preferred
Cholesterol Water Soluble is a major sterol in mammals, constituting 20-25% of the structural composition of the plasma membrane. It plays an important role in determining the fluidity and permeability characteristics of membranes and the function of transporters and signaling proteins. It is also an endogenous estrogen-related receptor alpha (ERRα) agonist. This product is a mixture of Cholesterol and Methyl-β-cyclodextrin, with specifications indicating only the effective content of Cholesterol.
  • Major sterol in mammals (20-25% of plasma membrane structural composition).
  • Plays a role in determining membrane fluidity and permeability.
  • Influences the function of transporters and signaling proteins.
  • Endogenous estrogen-related receptor alpha (ERRα) agonist.
  • Useful for studying cholesterol effects on potassium currents in inner hair cells.
  • Purity: 98.0%.
Catalog No. 50-004-27309 Supplier Medchemexpress LLC Supplier No. HYN0322A5MG
Cholesterol Water Soluble is a major sterol in mammals, constituting 20-25% of the structural composition of the plasma membrane. It plays an important role in determining the fluidity and permeability characteristics of membranes and the function of transporters and signaling proteins. It is also an endogenous estrogen-related receptor alpha (ERRα) agonist. This product is a mixture of Cholesterol and Methyl-β-cyclodextrin, with specifications indicating only the effective content of Cholesterol.
  • Major sterol in mammals (20-25% of plasma membrane structural composition).
  • Plays a role in determining membrane fluidity and permeability.
  • Influences the function of transporters and signaling proteins.
  • Endogenous estrogen-related receptor alpha (ERRα) agonist.
  • Useful for studying cholesterol effects on potassium currents in inner hair cells.
  • Purity: 98.0%.