Learn More
Cayman Chemical Cholesterol (solution), Purity: ≥95%, CAS: 57-88-5, 1 mL
SDP
Catalog No. 502918621
Encompass_Preferred
A sterol; a component of mammalian cell membranes that interacts with membrane phospholipids, sphingolipids, and proteins; a component of various LBDD systems, including liposomes and LNPs, where it has a role in membrane stability; a precursor of steroid hormones, bile acids, and the active form of vitamin D
Catalog No. 50-291-8621 Supplier Cayman Chemical Supplier No. 390881 ML
A sterol; a component of mammalian cell membranes that interacts with membrane phospholipids, sphingolipids, and proteins; a component of various LBDD systems, including liposomes and LNPs, where it has a role in membrane stability; a precursor of steroid hormones, bile acids, and the active form of vitamin D