Learn More
Cayman Chemical Cholesteryl Acetate 5g
SDP
Catalog No. 502989824
Encompass_Preferred
A cholesterol ester; has been used as an internal standard for the quantification of cholesterol, as well as cholesterol derivatives, by GC or GLC; has been derivatized from, and used for the quantification of, cholesterol by ESI-MS/MS
Catalog No. 50-298-9824 Supplier Cayman Chemical Supplier No. 308165 G
A cholesterol ester; has been used as an internal standard for the quantification of cholesterol, as well as cholesterol derivatives, by GC or GLC; has been derivatized from, and used for the quantification of, cholesterol by ESI-MS/MS