Learn More
Cayman Chemical Cholesteryl Arachidate, Purity: ≥98%, CAS: 2573-03-7, 100 mg
SDP
Catalog No. 502935410
Encompass_Preferred
A cholesterol ester found in human meibum; has been used as an internal standard for the quantification of fatty acids in B. napus
Catalog No. 50-293-5410 Supplier Cayman Chemical Supplier No. 26465100 MG
A cholesterol ester found in human meibum; has been used as an internal standard for the quantification of fatty acids in B. napus