Learn More
Cayman Chemical Cholesteryl ArachIdate 250mg
SDP
Catalog No. 502933455
Encompass_Preferred
A cholesterol ester found in human meibum; has been used as an internal standard for the quantification of fatty acids in B. napus
Catalog No. 50-293-3455 Supplier Cayman Chemical Supplier No. 26465250 MG
Add to Cart
Add to Cart
A cholesterol ester found in human meibum; has been used as an internal standard for the quantification of fatty acids in B. napus