Learn More
Cayman Chemical Cholesteryl Arachidate, Purity: ≥98%, CAS: 2573-03-7, 500 mg
SDP
Catalog No. 502934008
Encompass_Preferred
A cholesterol ester found in human meibum; has been used as an internal standard for the quantification of fatty acids in B. napus
Catalog No. 50-293-4008 Supplier Cayman Chemical Supplier No. 26465500 MG
A cholesterol ester found in human meibum; has been used as an internal standard for the quantification of fatty acids in B. napus