Learn More
Cayman Chemical Cholesteryl Arachidonate, Purity: ≥98%, CAS: 604-34-2, 25 mg
SDP
Catalog No. 502986698
Encompass_Preferred
A cholesterol ester; a component of LDL; oxidiation of the arachidonate moiety contributes contributes to macrophage activation and foam cell formation in atherosclerosis; undergoes hydrolysis to release arachidonic acid during ACTH-stimulated prostaglandin synthesis in rat adrenocortical cells; levels increased in BLAF in pediatric cycstic fibrosis patients and placentas of women with chorioamnionitis
Catalog No. 50-298-6698 Supplier Cayman Chemical Supplier No. 2259525 MG
A cholesterol ester; a component of LDL; oxidiation of the arachidonate moiety contributes contributes to macrophage activation and foam cell formation in atherosclerosis; undergoes hydrolysis to release arachidonic acid during ACTH-stimulated prostaglandin synthesis in rat adrenocortical cells; levels increased in BLAF in pediatric cycstic fibrosis patients and placentas of women with chorioamnionitis