Learn More
Cayman Chemical Cholesteryl Behenate, Purity: ≥98%, CAS: 61510-09-6, 250 mg
SDP
Catalog No. 502933456
Encompass_Preferred
A cholesterol ester; has been used as an internal standard for the quantification of cholesterol esters in human meibum samples
Catalog No. 50-293-3456 Supplier Cayman Chemical Supplier No. 26466250 MG
A cholesterol ester; has been used as an internal standard for the quantification of cholesterol esters in human meibum samples