Learn More
Cayman Chemical Cholesteryl Butyrate 25g
SDP
Catalog No. 502945079
Encompass_Preferred
A cholesterol ester; has been used with other lipids in the generation of LNPs; LNPs containing cholesteryl butyrate decrease proliferation, induce cell death, as well as decrease c-Myc levels in cancer cells
Catalog No. 50-294-5079 Supplier Cayman Chemical Supplier No. 4095025 G
A cholesterol ester; has been used with other lipids in the generation of LNPs; LNPs containing cholesteryl butyrate decrease proliferation, induce cell death, as well as decrease c-Myc levels in cancer cells