Learn More
Cayman Chemical Cholesteryl Butyrate, Purity: ≥95%, CAS: 521-13-1, 5 g
SDP
Catalog No. 502944817
Encompass_Preferred
A cholesterol ester; has been used with other lipids in the generation of LNPs; LNPs containing cholesteryl butyrate decrease proliferation, induce cell death, as well as decrease c-Myc levels in cancer cells
Catalog No. 50-294-4817 Supplier Cayman Chemical Supplier No. 409505 G
A cholesterol ester; has been used with other lipids in the generation of LNPs; LNPs containing cholesteryl butyrate decrease proliferation, induce cell death, as well as decrease c-Myc levels in cancer cells