Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl Chloride 98%, Thermo Scientific™

Catalog No. AC110210250
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC110210250 25 g
AC110210100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110210250 Supplier Thermo Scientific Chemicals Supplier No. 110210250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 910-31-6
Molecular Formula C27H45Cl
Molecular Weight (g/mol) 405.107
InChI Key OTVRYZXVVMZHHW-DGPGEHMFSA-N
Synonym cholesterylchloride
PubChem CID 131632519
IUPAC Name (8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C

Specifications

Melting Point 95°C to 97°C
Color White
Loss on Drying 0.2% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Quantity 25 g
Merck Index 09, 2194
Specific Rotation -22° to -26° (25°C, 589nm) (c=1, CHCl3)
Formula Weight 405.09
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Cholesteryl chloride, 98%
Show More Show Less
EINECSNumber 213-004-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.