Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAA1043314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1043314 25 g
AAA1043322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1043314 Supplier Thermo Scientific Chemicals Supplier No. A1043314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 910-31-6
Molecular Formula C27H45Cl
Molecular Weight (g/mol) 405.107
MDL Number MFCD00003632
InChI Key OTVRYZXVVMZHHW-DGPGEHMFSA-N
Synonym cholesterylchloride
PubChem CID 131632519
IUPAC Name (8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C

Specifications

Melting Point 94°C to 96°C
Quantity 25 g
Beilstein 2703655
Optical Rotation −26° (c=1 in benzene)
Formula Weight 405.11
Percent Purity 98%
Chemical Name or Material Cholesteryl chloride
EINECSNumber 213-004-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.