Learn More
Cayman Chemical Cholesteryl-d7 Palmitate, Purity: ≥99% deuterated forms (d1-d7), CAS: 1416275-32-5, 10 mg
SDP
Catalog No. 502893963
Encompass_Preferred
An internal standard for the quantification of cholesteryl palmitate by GC- or LC-MS
Catalog No. 50-289-3963 Supplier Cayman Chemical Supplier No. 900042210 MG
An internal standard for the quantification of cholesteryl palmitate by GC- or LC-MS