Learn More
Cayman Chemical Cholesteryl-d7 Palmitate, Purity: ≥99% deuterated forms (d1-d7), CAS: 1416275-32-5, 5 mg
SDP
Catalog No. 502893672
Encompass_Preferred
An internal standard for the quantification of cholesteryl palmitate by GC- or LC-MS
Catalog No. 50-289-3672 Supplier Cayman Chemical Supplier No. 90004225 MG
An internal standard for the quantification of cholesteryl palmitate by GC- or LC-MS