Learn More
Cayman Chemical Cholesteryl Eicosapentaenoate, Purity: ≥95%, CAS: 74892-97-0, 1 mg
SDP
Catalog No. 502994871
Encompass_Preferred
A cholesterol ester; induces cytotoxicity in human monocyte-macrophages at 88-880 μM; levels are elevated 1.89-fold in the serum of pediatric patients with extrahepatic biliary atresia and decreased in the serum and plasma of patients with Alzheimer’s disease and abetalipoproteinemia, respectively, compared with healthy individuals
Catalog No. 50-299-4871 Supplier Cayman Chemical Supplier No. 227191 MG
A cholesterol ester; induces cytotoxicity in human monocyte-macrophages at 88-880 μM; levels are elevated 1.89-fold in the serum of pediatric patients with extrahepatic biliary atresia and decreased in the serum and plasma of patients with Alzheimer’s disease and abetalipoproteinemia, respectively, compared with healthy individuals