Learn More
Cayman Chemical Cholesteryl Erucate 100mg
SDP
Catalog No. 502933617
Encompass_Preferred
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions
Catalog No. 50-293-3617 Supplier Cayman Chemical Supplier No. 26468100 MG
Add to Cart
Add to Cart
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions