Learn More
Cayman Chemical Cholesteryl Erucate, Purity: ≥98%, CAS: 24516-39-0, 250 mg
SDP
Catalog No. 502936905
Encompass_Preferred
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions
Catalog No. 50-293-6905 Supplier Cayman Chemical Supplier No. 26468250 MG
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions