Learn More
Cayman Chemical Cholesteryl Erucate, Purity: ≥98%, CAS: 24516-39-0, 500 mg
SDP
Catalog No. 502935447
Encompass_Preferred
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions
Catalog No. 50-293-5447 Supplier Cayman Chemical Supplier No. 26468500 MG
A cholesterol ester; has been used as a standard for the quantification of cholesterol esters in human meibomian gland secretions