Learn More
Kyfora Bio CHEMS (Cholesteryl hemisuccinate) 1 GR
Supplier:  Kyfora Bio 239671
Encompass_Preferred
Cholesteryl hemisuccinate (CHEMS) is a cholesterol derivate used in liposomal formulations that is responsive to changes in pH. CHEMS is negatively charged at neutral pH, but neutral at acidic pH, causing bilayer disruption. It can be used in acid-sensitive delivery systems for controlled drug release.Key application(s): liposome, acid-delivery systems
Catalog No. 50-255-9788