Learn More
Cayman Chemical Cholesteryl HemIsuccInate 5g
SDP
Catalog No. 502936478
Encompass_Preferred
A cholesterol ester with anticancer activity; inhibits the growth of murine C1498 myeloid and L1210 lymphocytic leukemia cells at 50 and 150 μM, respectively; an ionizable anionic detergent commonly used to stabilize unilamellar vesicles and liposomes; has been used as an emulsifying agent in various vesicular drug delivery systems for anticancer drugs, antibiotics, and oligonucleotides and to solubilize various proteins as well as erythrocyte ghosts
Catalog No. 50-293-6478 Supplier Cayman Chemical Supplier No. 256985 G
A cholesterol ester with anticancer activity; inhibits the growth of murine C1498 myeloid and L1210 lymphocytic leukemia cells at 50 and 150 μM, respectively; an ionizable anionic detergent commonly used to stabilize unilamellar vesicles and liposomes; has been used as an emulsifying agent in various vesicular drug delivery systems for anticancer drugs, antibiotics, and oligonucleotides and to solubilize various proteins as well as erythrocyte ghosts