Learn More
Cayman Chemical Cholesteryl Heptadecanoate, Purity: ≥95%, CAS: 24365-37-5, 500 mg
SDP
Catalog No. 502987375
Encompass_Preferred
A C-17 cholesterol ester commonly used as an internal standard for quantification of CEs by GC- or LC-mass spectrometry
Catalog No. 50-298-7375 Supplier Cayman Chemical Supplier No. 21750500 MG
A C-17 cholesterol ester commonly used as an internal standard for quantification of CEs by GC- or LC-mass spectrometry