Learn More
Cayman Chemical Cholesteryl Heptadecanoate, Purity: ≥95%, CAS: 24365-37-5, 1 g
SDP
Catalog No. 502985969
Encompass_Preferred
A C-17 cholesterol ester commonly used as an internal standard for quantification of CEs by GC- or LC-mass spectrometry
Catalog No. 50-298-5969 Supplier Cayman Chemical Supplier No. 217501 G
A C-17 cholesterol ester commonly used as an internal standard for quantification of CEs by GC- or LC-mass spectrometry