Learn More
Cayman Chemical Cholesteryl Heptadecanoate, Purity: ≥95%, CAS: 24365-37-5, 50 mg
SDP
Catalog No. 502987361
Encompass_Preferred
A C-17 cholesterol ester commonly used as an internal standard for quantification of CEs by GC- or LC-mass spectrometry
Catalog No. 50-298-7361 Supplier Cayman Chemical Supplier No. 2175050 MG
A C-17 cholesterol ester commonly used as an internal standard for quantification of CEs by GC- or LC-mass spectrometry